Tisk…

4,6,8-Trimethylquinoline 95%

Dodavatel: Apollo Scientific

OR937011-1G OR937011-250MG
APOSOR937011-1GEA 7580 CZK
APOSOR937011-1G APOSOR937011-250MG
4,6,8-Trimethylquinoline 95%
4,6,8-Trimethylquinoline
Vzorec: C₁₂H₁₃N
MW: 171,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191505
CAS číslo: 88565-88-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR