5,5-Dimethylacetylacetone 95%
Dodavatel: Apollo Scientific
Synonyms:
5,5-Dimethylhexane-2,4-dione, 2,2-Dimethyl-3,5-hexanedione, 5,5-Diméthylhexanedione-2,4
OR936943-1G
OR936943-5G
APOSOR936943-1GEA
4570
CZK
APOSOR936943-1G
APOSOR936943-5G
5,5-Dimethylacetylacetone 95%
5,5-Dimethylacetylacetone
Vzorec:
C₈H₁₄O₂ MW: 142,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00142767 CAS číslo: 7307-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...