Tisk…

4-Amino-2-methylquinoline 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Aminoquinaldine, 2-Methylquinolin-4-amine

OR936906-25G OR936906-5G
APOSOR936906-25GEA 5790 CZK
APOSOR936906-25G APOSOR936906-5G
4-Amino-2-methylquinoline 95%
4-Amino-2-methylquinoline
Vzorec: C₁₀H₁₀N₂
MW: 158,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006759
CAS číslo: 6628-04-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR