Tisk…

N-Cyanomethyl-N-methyl-4-nitroaniline 95%

Dodavatel: Apollo Scientific

OR936889-1G OR936889-5G
APOSOR936889-1GEA 5590 CZK
APOSOR936889-1G APOSOR936889-5G
N-Cyanomethyl-N-methyl-4-nitroaniline 95%
N-Cyanomethyl-N-methyl-4-nitroaniline
Vzorec: C₉H₉N₃O₂
MW: 191.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191337
CAS číslo: 107023-66-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR