Tisk…

2-Hydroxyethyl-4-hydroxybenzoate 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Hydroxybenzoic Acid 2-Hydroxyethyl Ester, 2-Hydroxyethylparaben

OR936885-100G OR936885-10G OR936885-25G
APOSOR936885-100GEA 20900 CZK
APOSOR936885-100G APOSOR936885-10G APOSOR936885-25G
2-Hydroxyethyl-4-hydroxybenzoate 95%
2-Hydroxyethyl-4-hydroxybenzoate
Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059079
CAS číslo: 2496-90-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR