2-(4-tert-Butylphenyl)ethanol 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(4-(tert-Butyl)phenyl)ethanol
OR936878-10G
OR936878-25G
OR936878-5G
APOSOR936878-10GEA
2110
CZK
APOSOR936878-10G
APOSOR936878-25G
APOSOR936878-5G
2-(4-tert-Butylphenyl)ethanol 95%
2-(4-tert-Butylphenyl)ethanol
Vzorec:
C₁₂H₁₈O Storage Temperature: Ambient |
MDL Number:
MFCD00236023 CAS číslo: 5406-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...