Tisk…

2-Propionylpyrrole 95%

Dodavatel: Apollo Scientific

OR936803-1G OR936803-25G OR936803-5G
APOSOR936803-1GEA 1240 CZK
APOSOR936803-1G APOSOR936803-25G APOSOR936803-5G
2-Propionylpyrrole 95%
2-Propionylpyrrole
Vzorec: C₇H₉NO
Storage Temperature: Ambient
MDL Number: MFCD01696449
CAS číslo: 1073-26-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR