trans-2-Methyl-2-pentenoic acid 95%
Dodavatel: Apollo Scientific
Synonyms:
(E)-2-Methylpent-2-enoic acid
OR936754-100G
OR936754-25G
APOSOR936754-100GEA
8270
CZK
APOSOR936754-100G
APOSOR936754-25G
trans-2-Methyl-2-pentenoic acid 95%
trans-2-Methyl-2-pentenoic acid
Vzorec:
C₆H₁₀O₂ MW: 114,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002655 CAS číslo: 16957-70-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...