Tisk…

tert-Butyldimethylsilyl-N-phenylbenzimidate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: O-(tert-Butyldimethylsilyl)benzanilide, TBDMS-BEZA

OR936738-1G OR936738-5G
APOSOR936738-1GEA 1870 CZK
APOSOR936738-1G APOSOR936738-5G
tert-Butyldimethylsilyl-N-phenylbenzimidate 98%
tert-Butyldimethylsilyl-N-phenylbenzimidate
Vzorec: C₁₉H₂₅NOSi
MW: 311,50 g/mol
Bod tání: 71 °C
Storage Temperature: Ambient
CAS číslo: 404392-70-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR