Tisk…

1-Cyclohexylethanol 95%

Dodavatel: Apollo Scientific

OR936737-25G OR936737-5G
APOSOR936737-25GEA 7080 CZK
APOSOR936737-25G APOSOR936737-5G
1-Cyclohexylethanol 95%
1-Cyclohexylethanol
Vzorec: C₈H₁₆O
MW: 128.21
Hustota: 0.918
Storage Temperature: Ambient
MDL Number: MFCD00001475
CAS číslo: 1193-81-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR