3,4-Dimethylphenyl isocyanate 95%
Dodavatel: Apollo Scientific
Synonyms:
3,4-Xylyl isocyanate, 4-Isocyanato-1,2-dimethylbenzene, 3,4-Dimethylphenylisocyanate
OR936712-25G
APOSOR936712-25GEA
3150
CZK
APOSOR936712-25G
3,4-Dimethylphenyl isocyanate 95%
3,4-Dimethylphenyl isocyanate
- May form small amount of sediment.
Vzorec:
C₉H₉NO MW: 147.18 Storage Temperature: Ambient |
MDL Number:
MFCD00013867 CAS číslo: 51163-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...