Tisk…

Phenyl acrylate 95%

Dodavatel: Apollo Scientific

OR936700-25G
APOSOR936700-25GEA 3310 CZK
APOSOR936700-25G
Phenyl acrylate 95%
Phenyl acrylate
Vzorec: C₉H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00048145
CAS číslo: 937-41-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR