Tisk…

N-Acetyl-ε-caprolactam 95%

Dodavatel: Apollo Scientific

Synonyms: N-Acetyl-2-oxohexamethyleneimine, N-Acetylcaprolactam

OR936673-500G
APOSOR936673-500GEA 3190 CZK
APOSOR936673-500G
N-Acetyl-ε-caprolactam 95%
N-Acetyl-epsilon-caprolactam
Vzorec: C₈H₁₃NO₂
MW: 155,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003262
CAS číslo: 1888-91-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR