3-Benzyloxazolidine 95%
Dodavatel: Apollo Scientific
OR936664-1G
OR936664-5G
APOSOR936664-1GEA
2400
CZK
APOSOR936664-1G
APOSOR936664-5G
3-Benzyloxazolidine 95%
3-Benzyloxazolidine
Vzorec:
C₁₀H₁₃NO MW: 163,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059028 CAS číslo: 13657-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...