Tisk…

1-Methylbenzotriazole 95%

Dodavatel: Apollo Scientific

Synonyms: 1-Methyl-1H-benzotriazole

OR936661-25G OR936661-5G
APOSOR936661-25GEA 5460 CZK
APOSOR936661-25G APOSOR936661-5G
1-Methylbenzotriazole 95%
1-Methylbenzotriazole
Vzorec: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD00014572
CAS číslo: 13351-73-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR