Ethyl cycloheptylideneacetate 95%
Dodavatel: Apollo Scientific
OR936634-1G
OR936634-5G
APOSOR936634-1GEA
2860
CZK
APOSOR936634-1G
APOSOR936634-5G
Ethyl cycloheptylideneacetate 95%
Ethyl cycloheptylideneacetate
Vzorec:
C₁₁H₁₈O₂ Storage Temperature: Ambient |
MDL Number:
MFCD02093941 CAS číslo: 1903-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...