Tisk…
Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers) 95%
Dodavatel: Apollo Scientific
Synonyms: Ethyl 3-Methyl-5-cyclohexen-1-one-2-carboxylate, 6-Methyl-2-oxo-3-cyclohexene-1-carboxylic Acid Ethyl Ester
OR936598-10G OR936598-1G OR936598-5G
APOSOR936598-10GEA 19840 CZK
APOSOR936598-10G APOSOR936598-1G APOSOR936598-5G
Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers) 95%
Ethyl-6-methyl-2-oxo-3-cyclohexene-1-carboxylate (mixture of isomers)
Vzorec: C₁₀H₁₄O₃
MW: 182.22 g/mol
Storage Temperature: Ambient
CAS číslo: 3419-32-7