trans-4-tert-Butylcyclohexanol 95%
Dodavatel: Apollo Scientific
Synonyms:
trans-4-(tert-Butyl)cyclohexanol
OR936549-1G
OR936549-250MG
OR936549-5G
APOSOR936549-1GEA
3610
CZK
APOSOR936549-1G
APOSOR936549-250MG
APOSOR936549-5G
trans-4-tert-Butylcyclohexanol 95%
trans-4-tert-Butylcyclohexanol
Vzorec:
C₁₀H₂₀O MW: 156,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00064952 CAS číslo: 21862-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...