Tisk…

1,1-Diphenylhydrazine 95%

Dodavatel: Apollo Scientific

OR936542-10G OR936542-1G OR936542-5G
APOSOR936542-10GEA 22750 CZK
APOSOR936542-10G APOSOR936542-1G APOSOR936542-5G
1,1-Diphenylhydrazine 95%
1,1-Diphenylhydrazine
Vzorec: C₁₂H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00044505
CAS číslo: 530-50-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR