Tisk…

2-Hydroxy-N-methylbenzylamine hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: N-Methyl-2-hydroxybenzylamine Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, alpha-Methylamino-o-cresol Hydrochloride

OR936532-1G OR936532-5G OR936532-25G
APOSOR936532-1GEA 2440 CZK
APOSOR936532-1G APOSOR936532-5G APOSOR936532-25G
2-Hydroxy-N-methylbenzylamine hydrochloride 95%
2-Hydroxy-N-methylbenzylamine hydrochloride
Vzorec: C₈H₁₂NO
Storage Temperature: Ambient
MDL Number: MFCD00142934
CAS číslo: 60399-02-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR