2-Methylbenzothiazol-6-ol 97%
Dodavatel: Apollo Scientific
Synonyms:
2-Methylbenzo[d]thiazol-6-ol
OR936497-1G
OR936497-5G
APOSOR936497-1GEA
1040
CZK
APOSOR936497-1G
APOSOR936497-5G
2-Methylbenzothiazol-6-ol 97%
2-Methylbenzothiazol-6-ol
Vzorec:
C₈H₇NOS MW: 165,21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00227234 CAS číslo: 68867-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...