Tisk…

3-Chloropropyl isocyanate 95%

Dodavatel: Apollo Scientific

OR936473-1G OR936473-5G
APOSOR936473-1GEA 3740 CZK
APOSOR936473-1G APOSOR936473-5G
3-Chloropropyl isocyanate 95%
3-Chloropropyl isocyanate

  • May form small amount of sediment.

Vzorec: C₄H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD00010384
CAS číslo: 13010-19-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR