Tisk…

(R)-(+)-2-Chlorobutyric acid 95%

Dodavatel: Apollo Scientific

OR936459-1G OR936459-250MG
APOSOR936459-1GEA 10150 CZK
APOSOR936459-1G APOSOR936459-250MG
(R)-(+)-2-Chlorobutyric acid 95%
(R)-(+)-2-Chlorobutyric acid
Vzorec: C₄H₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00067098
CAS číslo: 54053-45-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR