Tisk…

1,1-Bis(morpholino)ethylene 95%

Dodavatel: Apollo Scientific

OR936449-10G OR936449-1G OR936449-5G
APOSOR936449-10GEA 21730 CZK
APOSOR936449-10G APOSOR936449-1G APOSOR936449-5G
1,1-Bis(morpholino)ethylene 95%
1,1-Bis(morpholino)ethylene
Vzorec: C₁₀H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00059775
CAS číslo: 14212-87-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR