Tisk…

2-Biphenylylmethanol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Biphenylmethanol, 2-Phenylbenzyl alcohol, [1,1-Biphenyl]-2-ylmethanol

OR936438-1G OR936438-5G
APOSOR936438-1GEA 1210 CZK
APOSOR936438-1G APOSOR936438-5G
2-Biphenylylmethanol 95%
2-Biphenylylmethanol
Vzorec: C₁₃H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00004620
CAS číslo: 2928-43-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR