4'-Methylpropiophenone 95%
Dodavatel: Apollo Scientific
Synonyms:
p-Methylpropiophenone
OR936436-500G
APOSOR936436-500GEA
1740
CZK
APOSOR936436-500G
4'-Methylpropiophenone 95%
4'-Methylpropiophenone
Vzorec:
C₁₀H₁₂O MW: 148,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009312 CAS číslo: 5337-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...