tert-Butylacetyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,3-Dimethylbutyryl chloride
OR936433-100G
OR936433-25G
APOSOR936433-100GEA
16960
CZK
APOSOR936433-100G
APOSOR936433-25G
tert-Butylacetyl chloride 95%
tert-Butylacetyl chloride
Vzorec:
C₆H₁₁ClO MW: 134.61 g/mol Bod varu: 127…129 °C Hustota: 0.969 g/cm³ (25 °C) Bod vzplanutí: 21 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000737 CAS číslo: 7065-46-5 UN: 2924 ADR: 3+8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...