Methyl-5-chloro-5-oxovalerate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl glutaryl chloride, Methyl 4-chloroformylbutyrate
OR936413-25G
APOSOR936413-25GEA
3940
CZK
APOSOR936413-25G
Methyl-5-chloro-5-oxovalerate 95%
Methyl-5-chloro-5-oxovalerate
Vzorec:
C₆H₉ClO₃ MW: 164,59 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00000756 CAS číslo: 1501-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...