2-Methylcyclopropanecarboxylic acid 96%
Dodavatel: Apollo Scientific
OR936381-10G
OR936381-1G
OR936381-5G
APOSOR936381-10GEA
8600
CZK
APOSOR936381-10G
APOSOR936381-1G
APOSOR936381-5G
2-Methylcyclopropanecarboxylic acid 96%
2-Methylcyclopropanecarboxylic acid
Vzorec:
C₅H₈O₂ MW: 100.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00001293 CAS číslo: 29555-02-0 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...