Tisk…

2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene 95%

Dodavatel: Apollo Scientific

OR936368-1G OR936368-25G OR936368-5G
APOSOR936368-1GEA 2900 CZK
APOSOR936368-1G APOSOR936368-25G APOSOR936368-5G
2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene 95%
2,4,4,6-Tetramethyl-1-oxa-3-aza-2-cyclohexene
Vzorec: C₈H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD00014605
CAS číslo: 26939-18-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR