Tisk…

4-Hydroxy-3,5-dimethylbenzoic acid 97%

Dodavatel: Apollo Scientific

OR936362-25G
APOSOR936362-25GEA 5130 CZK
APOSOR936362-25G
4-Hydroxy-3,5-dimethylbenzoic acid 97%
4-Hydroxy-3,5-dimethylbenzoic acid
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016536
CAS číslo: 4919-37-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR