Tisk…

(±)-α-Terpineol 95%

Dodavatel: Apollo Scientific

OR936354-100G OR936354-500G
APOSOR936354-100GEA 870 CZK
APOSOR936354-100G APOSOR936354-500G
(±)-α-Terpineol 95%
(±)-α-Terpineol
Vzorec: C₁₀H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD00001557
CAS číslo: 98-55-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR