4,4'-Dipyridyl sulphide 95%
Dodavatel: Apollo Scientific
Synonyms:
4,4'-Thiodipyridine, Di(pyridin-4-yl)sulphane
OR936342-1G
OR936342-5G
APOSOR936342-1GEA
2730
CZK
APOSOR936342-1G
APOSOR936342-5G
4,4'-Dipyridyl sulphide 95%
4,4'-Dipyridyl sulphide
Vzorec:
C₁₀H₈N₂S Storage Temperature: Ambient |
MDL Number:
MFCD00059784 CAS číslo: 37968-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...