Tisk…

2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene 95%

Dodavatel: Apollo Scientific

OR936341-1G OR936341-250MG
APOSOR936341-1GEA 9330 CZK
APOSOR936341-1G APOSOR936341-250MG
2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene 95%
2,6-Bis(hydroxymethyl)-1,4-dimethoxybenzene
Vzorec: C₁₀H₁₄O₄
MW: 198,22 g/mol
Storage Temperature: Ambient
CAS číslo: 78840-04-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR