Tisk…

2-Cyclohexylphenol 95%

Dodavatel: Apollo Scientific

OR936286-1G OR936286-25G OR936286-5G
APOSOR936286-1GEA 0 CZK
APOSOR936286-1G APOSOR936286-25G APOSOR936286-5G
2-Cyclohexylphenol 95%
2-Cyclohexylphenol
Vzorec: C₁₂H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD00019335
CAS číslo: 119-42-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR