Tisk…

2-Ethyl-3-methoxypyrazine 95%

Dodavatel: Apollo Scientific

OR936279-25G
APOSOR936279-25GEA 3230 CZK
APOSOR936279-25G
2-Ethyl-3-methoxypyrazine 95%
2-Ethyl-3-methoxypyrazine
Vzorec: C₇H₁₀N₂O
MW: 138,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006129
CAS číslo: 25680-58-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR