Tisk…

5-Phenyl-4-pentyn-1-ol 95%

Dodavatel: Apollo Scientific

Synonyms: 5-Phenylpent-4-yn-1-ol

OR936269-1G OR936269-250MG
APOSOR936269-1GEA 6090 CZK
APOSOR936269-1G APOSOR936269-250MG
5-Phenyl-4-pentyn-1-ol 95%
5-Phenyl-4-pentyn-1-ol
Vzorec: C₁₁H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD00191481
CAS číslo: 24595-58-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR