Tisk…

4-Propoxyaniline 98%

Dodavatel: Apollo Scientific

OR936244-25G OR936244-5G
APOSOR936244-25GEA 13860 CZK
APOSOR936244-25G APOSOR936244-5G
4-Propoxyaniline 98%
4-Propoxyaniline
Vzorec: C₉H₁₃NO
MW: 151.21
Storage Temperature: Ambient
MDL Number: MFCD00043621
CAS číslo: 4469-80-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR