Tisk…

Ethyl isopropyl sulphone 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(Ethylsulphonyl)propane

OR936236-25G
APOSOR936236-25GEA 2780 CZK
APOSOR936236-25G
Ethyl isopropyl sulphone 95%
Ethyl isopropyl sulphone
Vzorec: C₅H₁₂O₂S
MW: 136,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050418
CAS číslo: 4853-75-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR