Tisk…

2-Acetyl-7-hydroxybenzofuran 95%

Dodavatel: Apollo Scientific

OR936224-10G OR936224-1G OR936224-5G
APOSOR936224-10GEA 4270 CZK
APOSOR936224-10G APOSOR936224-1G APOSOR936224-5G
2-Acetyl-7-hydroxybenzofuran 95%
2-Acetyl-7-hydroxybenzofuran
Vzorec: C₁₀H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00191373
CAS číslo: 40020-87-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR