2-Bromo-3-pyridineboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromopyridine-3-boronic acid, (2-Bromopyridin-3-yl)boronic acid, 2-Bromo-3-pyridinylboronic acid
OR9358-25G
APOSOR9358-25GEA
5200
CZK
APOSOR9358-25G
2-Bromo-3-pyridineboronic acid
2-Bromo-3-pyridineboronic acid
Vzorec:
C₅H₅BBrNO₂ MW: 201,81 g/mol Bod tání: 131…132 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD04113805 CAS číslo: 452972-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...