Tisk…

5-Chloro-2-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

OR9343-1G OR9343-5G
APOSOR9343-1GEA 3940 CZK
APOSOR9343-1G APOSOR9343-5G
5-Chloro-2-pyridineboronic acid
5-Chloro-2-pyridineboronic acid
Vzorec: C₅H₅BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD07368862
CAS číslo: 652148-91-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR