Tisk…

4-Methylveratrole 95%

Dodavatel: Apollo Scientific

Synonyms: 1,2-Dimethoxy-4-methylbenzene, 3,4-Dimethoxytoluene

OR933361-500G
APOSOR933361-500GEA 3640 CZK
APOSOR933361-500G
4-Methylveratrole 95%
4-Methylveratrole
Vzorec: C₉H₁₂O₂
MW: 152,19 g/mol
Bod varu: 133…135 °C/50mmHg
Bod tání: 22…23 °C
Hustota: 1,0509 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016651
CAS číslo: 494-99-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR