Tisk…

H-D-Nle-OMe·HCl 98%

Dodavatel: Apollo Scientific

Synonyms: D-Norleucine methyl ester hydrochloride

OR933329-10G OR933329-1G OR933329-5G
APOSOR933329-10GEA 16340 CZK
APOSOR933329-10G APOSOR933329-1G APOSOR933329-5G
H-D-Nle-OMe·HCl 98%
H-D-Nle-OMe·HCl
Vzorec: C₇H₁₅NO₂·HCl
Storage Temperature: Ambient
CAS číslo: 60687-33-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR