Tisk…

N-Carbobenzoxy-DL-norvaline 97%

Dodavatel: Apollo Scientific

OR933315-1G OR933315-25G OR933315-5G
APOSOR933315-1GEA 612 CZK
APOSOR933315-1G APOSOR933315-25G APOSOR933315-5G
N-Carbobenzoxy-DL-norvaline 97%
N-Carbobenzoxy-DL-norvaline
Vzorec: C₁₃H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD00020434
CAS číslo: 21691-43-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR