3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide 95%
Dodavatel: Apollo Scientific
OR933276-25G
OR933276-5G
APOSOR933276-25GEA
3480
CZK
APOSOR933276-25G
APOSOR933276-5G
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide 95%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)-N'-[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]propanehydrazide
Vzorec:
C₃₄H₅₂N₂O₄ MW: 552,80 g/mol Bod varu: 60…67 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00072662 CAS číslo: 32687-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...