Tisk…

Piperidine-1-sulphonyl chloride 97%

Dodavatel: Apollo Scientific

OR933169-25G
APOSOR933169-25GEA 8040 CZK
APOSOR933169-25G
Piperidine-1-sulphonyl chloride 97%
Piperidine-1-sulphonyl chloride
Vzorec: C₅H₁₀ClNO₂S
MW: 183,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03250329
CAS číslo: 35856-62-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR