Tisk…

4-Chloro-6-nitro-m-cresol 95%

Dodavatel: Apollo Scientific

OR933125-1G OR933125-25G OR933125-5G
APOSOR933125-1GEA 3740 CZK
APOSOR933125-1G APOSOR933125-25G APOSOR933125-5G
4-Chloro-6-nitro-m-cresol 95%
4-Chloro-6-nitro-m-cresol
Vzorec: C₇H₆ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00007114
CAS číslo: 7147-89-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR