Tisk…

tert-Butyl glycolate 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 2-hydroxyacetate

OR932881-100G
APOSOR932881-100GEA 4900 CZK
APOSOR932881-100G
tert-Butyl glycolate 95%
tert-Butyl glycolate
Vzorec: C₆H₁₂O₃
MW: 132,159 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03452739
CAS číslo: 50595-15-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR