Tisk…

4-Ethylbenzyl alcohol 98%

Dodavatel: Apollo Scientific

Synonyms: (4-Ethylphenyl)methanol

OR932622-25G OR932622-5G
APOSOR932622-25GEA 7870 CZK
APOSOR932622-25G APOSOR932622-5G
4-Ethylbenzyl alcohol 98%
4-Ethylbenzyl alcohol
Vzorec: C₉H₁₂O
MW: 136,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004666
CAS číslo: 768-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR